2-氯苯基乙醇 ≥97.0% (by GC)

供应商: TCI
Warning

C2169-1G C2169-5G
TCIAC2169-1GEA 0 CNY
TCIAC2169-1G TCIAC2169-5G
2-氯苯基乙醇 ≥97.0% (by GC)
2-氯苯基乙醇
分子式: C₈H₉ClO
分子量: 156.61 g/mol
沸点: 130°C
MDL编号: MFCD00002888
CAS号: 19819-95-5
EINECS: 243-348-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e9aaa1fbb1c02191a41b3133d54c8678dd68d56c&vid=e9aaa1fbb1c02191a41b3133d54c8678dd68d56c&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR