1-Bromo-4-propoxybenzene ≥98.0% (by GC)

供应商: TCI

Synonyms: 1-Bromo-4-n-propoxybenzene, 4-Propoxybromobenzene, 1-Bromo-4-propoxylbenzene

B3639-25G B3639-5G
TCIAB3639-25GEA 0 CNY
TCIAB3639-25G TCIAB3639-5G
1-Bromo-4-propoxybenzene ≥98.0% (by GC)
1-Bromo-4-propoxybenzene
分子式: C₉H₁₁BrO
分子量: 215.09 g/mol
沸点: 240°C
MDL编号: MFCD00156160
CAS号: 39969-56-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=517faa17ba40f602d1be59ba968f697d33419c75&vid=517faa17ba40f602d1be59ba968f697d33419c75&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR