2-溴-4-氟碘苯 ≥98.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
3-Bromo-1-fluoro-4-iodobenzene, 2-Bromo-4-fluoroiodobenzene, 1-Bromo-5-fluoro-2-iodobenzene
B3098-1G
B3098-5G
TCIAB3098-1GEA
0
CNY
TCIAB3098-1G
TCIAB3098-5G
2-溴-4-氟碘苯 ≥98.0% (by GC)
2-溴-4-氟碘苯
分子式:
C₆H₃BrFI 分子量: 300.9 g/mol |
MDL编号:
MFCD00142868 CAS号: 202865-73-4 |
|
产品技术规格
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...