4-溴-2-氟联苯 ≥99.0% (by GC)

供应商: TCI
Warning

Synonyms: 4-Bromo-2-fluoro-1,1'-biphenyl

B2028-25G B2028-250G
TCIAB2028-25GEA 0 CNY
TCIAB2028-25G TCIAB2028-250G
4-溴-2-氟联苯 ≥99.0% (by GC)
4-溴-2-氟联苯
分子式: C₁₂H₈BrF
分子量: 251.1 g/mol
沸点: 299
溶点: 38.5
MDL编号: MFCD00051716
CAS号: 41604-19-7
EINECS: 255-453-9
UN号: 3152
ADR: 9 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd5af3f6afe6a5d4c73084fa658686d505f75e5b&vid=fd5af3f6afe6a5d4c73084fa658686d505f75e5b&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Orange to Green powder to lump
Purity (GC) min. 99.0 %
Melting point 39.0 to 43.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR