1-氨基碘代吡啶 ≥98.0% (by titrimetric analysis)

供应商: TCI
Warning

Synonyms: 1-Aminopyridinium Iodide, 1-Aminopyridine hydroiodide, 1-Aminopyridine HI

A0629-25G A0629-5G
TCIAA0629-25GEA 0 CNY
TCIAA0629-25G TCIAA0629-5G
1-氨基碘代吡啶 ≥98.0% (by titrimetric analysis)
1-氨基碘代吡啶
分子式: C₅H₇IN₂
溶点: 161…162 °C
MDL编号: MFCD00051893
CAS号: 6295-87-0
EINECS: 228-566-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1dbacadaeb07d1a0c571cde19ff622d26253ff1d&vid=1dbacadaeb07d1a0c571cde19ff622d26253ff1d&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Light yellow to Light orange powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR