4-庚基苯胺 ≥98.0% (by GC, titration analysis)
供应商: TCI
|
Warning
|
H0588-25ML
H0588-5ML
TCIAH0588-25MLEA
0
CNY
TCIAH0588-25ML
TCIAH0588-5ML
4-庚基苯胺 ≥98.0% (by GC, titration analysis)
4-庚基苯胺
分子式:
C₁₃H₂₁N 分子量: 191.32 g/mol 沸点: 286°C |
MDL编号:
MFCD00007928 CAS号: 37529-27-4 |
|
产品技术规格
Appearance | Colorless to Yellow to Orange clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...