2-氨基苯乙醇 ≥95.0% (by GC, titration analysis)
供应商: TCI
|
Warning
|
Synonyms:
2-(2-Aminophenyl)ethyl alcohol, 2-Aminophenethyl alcohol
A2327-25G
A2327-5G
TCIAA2327-25GEA
0
CNY
TCIAA2327-25G
TCIAA2327-5G
2-氨基苯乙醇 ≥95.0% (by GC, titration analysis)
2-氨基苯乙醇
分子式:
C₈H₁₁NO 分子量: 137.18 g/mol 沸点: 148°C 溶点: 89 密度: 1.045 |
MDL编号:
MFCD00007754 CAS号: 5339-85-5 EINECS: 226-275-9 |
|
产品技术规格
Appearance | Light yellow to Yellow to Orange clear liquid |
Purity (GC) | min. 95.0 % |
Purity (Nonaqueous Titration) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...