N-氨基邻苯二甲酰亚胺 ≥98.0% (by HPLC)

供应商: TCI
Danger

Synonyms: 氨基邻苯二甲胺,不对称

A0843-25G A0843-5G
TCIAA0843-25GEA 0 CNY
TCIAA0843-25G TCIAA0843-5G
N-氨基邻苯二甲酰亚胺 ≥98.0% (by HPLC)
N-氨基邻苯二甲酰亚胺
分子式: C₈H₆N₂O₂
沸点: 391 °C (1013 hPa)
溶点: 203…206 °C
MDL编号: MFCD00005895
CAS号: 1875-48-5
EINECS: 217-505-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1df02721ac60172d6c968c4b3612b0c767520642&vid=1df02721ac60172d6c968c4b3612b0c767520642&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 16.50 to 18.00 %
Solubility in hot Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR