10-十一烯酸乙酯 ≥97.0% (by GC)
供应商: TCI
Synonyms:
十一烯酸乙酯, 10-十一烯酸乙酯
E0771-250G
E0771-25G
TCIAE0771-250GEA
0
CNY
TCIAE0771-250G
TCIAE0771-25G
10-十一烯酸乙酯 ≥97.0% (by GC)
10-十一烯酸乙酯
分子式:
C₁₃H₂₄O₂ 分子量: 212.33 g/mol 沸点: 259°C 溶点: –38°C |
MDL编号:
MFCD00009220 CAS号: 692-86-4 EINECS: 211-734-8 |
|
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...