9-癸烯-1-醇 ≥97.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
1-Decen-10-ol
D1892-25ML
D1892-500ML
TCIAD1892-25MLEA
0
CNY
TCIAD1892-25ML
TCIAD1892-500ML
9-癸烯-1-醇 ≥97.0% (by GC)
9-癸烯-1-醇
分子式:
C₁₀H₂₀O 分子量: 156.27 g/mol 沸点: 108°C 溶点: -13 闪点: 114°C |
MDL编号:
MFCD00002992 CAS号: 13019-22-2 EINECS: 235-878-6 |
|
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...