1,4-Diacetoxybenzene ≥98.0% (by GC)

供应商: TCI
Warning

Synonyms: Hydroquinone diacetate, 1,4-Phenylene diacetate, p-Phenylene di(acetate)

D1803-25G D1803-500G
TCIAD1803-25GEA 0 CNY
TCIAD1803-25G TCIAD1803-500G
1,4-Diacetoxybenzene ≥98.0% (by GC)
1,4-Diacetoxybenzene
分子式: C₁₀H₁₀O₄
沸点: 273 °C (1013 hPa)
溶点: 119…123 °C
MDL编号: MFCD00011643
CAS号: 1205-91-0
EINECS: 214-887-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2a4c25349f79752dc218b8f2f11d388205293afd&vid=2a4c25349f79752dc218b8f2f11d388205293afd&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 119.0 to 123.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR