1,2-Bis(trimethylsilyl)benzene ≥95.0% (by GC)
供应商: TCI
Synonyms:
1,2-Phenylenebis(trimethylsilane)
B2299-1G
B2299-5G
TCIAB2299-1GEA
0
CNY
TCIAB2299-1G
TCIAB2299-5G
1,2-Bis(trimethylsilyl)benzene ≥95.0% (by GC)
1,2-Bis(trimethylsilyl)benzene
分子式:
C₁₂H₂₂Si₂ 分子量: 222.48 g/mol 存储条件: Refrigerator |
MDL编号:
MFCD00015590 CAS号: 17151-09-6 |
|
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...