2-甲氧基苯乙腈 ≥98.0% (通过GC)

供应商: TCI

M0855-25G M0855-500G
TCIAM0855-25GEA 0 CNY
TCIAM0855-25G TCIAM0855-500G
2-甲氧基苯乙腈 ≥98.0% (通过GC)
2-甲氧基苯乙腈

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 67.0 to 71.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR