2-氟-6-(三氟甲基)溴苄 ≥98.0% (by GC)

供应商: TCI
Danger

Synonyms: 1-(Bromomethyl)-2-fluoro-6-(trifluoromethyl)benzene, 2-(Bromomethyl)-1-fluoro-3-(trifluoromethyl)benzene

F0732-1G F0732-5G
TCIAF0732-1GEA 0 CNY
TCIAF0732-1G TCIAF0732-5G
2-氟-6-(三氟甲基)溴苄 ≥98.0% (by GC)
2-氟-6-(三氟甲基)溴苄
分子式: C₈H₅BrF₄
分子量: 257.03 g/mol
沸点: 197
MDL编号: MFCD00082477
CAS号: 239087-08-2
UN号: 3261
ADR: 8 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b70f8a1e7f46387f5d98368d6e3a7ecbf83ead10&vid=b70f8a1e7f46387f5d98368d6e3a7ecbf83ead10&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 42.0 to 46.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR