苄基三丁基溴化铵 ≥98.0% (by titrimetric analysis)

供应商: TCI
Warning

B1217-25G B1217-500G
TCIAB1217-25GEA 0 CNY
TCIAB1217-25G TCIAB1217-500G
苄基三丁基溴化铵 ≥98.0% (by titrimetric analysis)
苄基三丁基溴化铵
分子式: C₁₉H₃₄BrN
MDL编号: MFCD00011848
CAS号: 25316-59-0
EINECS: 246-819-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d3bcc75f53c2955eb06208314d3d37547c5748b6&vid=d3bcc75f53c2955eb06208314d3d37547c5748b6&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR