N-甲基环己胺 ≥99.0% (by titrimetric analysis)
供应商: TCI
|
Danger
|
Synonyms:
N-环己基甲胺
M0202-25ML
M0202-500ML
TCIAM0202-25MLEA
0
CNY
TCIAM0202-25ML
TCIAM0202-500ML
N-甲基环己胺 ≥99.0% (by titrimetric analysis)
N-甲基环己胺
分子式:
C₇H₁₅N 沸点: 148°C 溶点: –8 °C 密度: 0,859 g/cm³ (20 °C) 闪点: 29°C |
MDL编号:
MFCD00003832 CAS号: 100-60-7 EINECS: 202-869-3 UN号: 2734 ADR: 8,II |
|
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...