原甲酸三乙酯 ≥98.0% (by GC)

供应商: TCI
Warning

Synonyms: 1,1,1-三乙氧基甲烷

O0066-25ML O0066-500ML
TCIAO0066-25MLEA 0 CNY
TCIAO0066-25ML TCIAO0066-500ML
原甲酸三乙酯 ≥98.0% (by GC)
原甲酸三乙酯
分子式: C₇H₁₆O₃
沸点: 145°C
溶点: –61 °C
密度: 0,8964 g/cm³ (20 °C)
闪点: 35°C
存储条件: Ambient
MDL编号: MFCD00009230
CAS号: 122-51-0
EINECS: 204-550-4
UN号: 2524
ADR: 3,III
Merck Index: 14,06884
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ac7722e46307866baa4a3c7268335d9e4d526551&vid=ac7722e46307866baa4a3c7268335d9e4d526551&prid=web&clid=SAPDAM

现在订购

产品技术规格

Purity (GC) min. 98.0 %
Appearance Colorless to Almost colorless clear liquid
Water max. 0.1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR