(溴甲基)三甲基硅烷 ≥97.0% (by GC)
供应商: TCI
|
Danger
|
Synonyms:
(甲基溴)三甲基硅烷
B1892-25G
B1892-5G
TCIAB1892-25GEA
0
CNY
TCIAB1892-25G
TCIAB1892-5G
(溴甲基)三甲基硅烷 ≥97.0% (by GC)
(溴甲基)三甲基硅烷
分子式:
BrH₂CSi(CH₃)₃ 沸点: 117°C 密度: 1,17 g/cm³ (25 °C) 闪点: 16°C |
MDL编号:
MFCD00000167 CAS号: 18243-41-9 UN号: 2924 ADR: 3,II |
|
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...