溴三苯基甲烷 ≥98.0% (by titrimetric analysis)
供应商: TCI
|
Danger
|
Synonyms:
三苯基溴甲烷, 三苯甲基溴
T0512-100G
T0512-25G
TCIAT0512-100GEA
0
CNY
TCIAT0512-100G
TCIAT0512-25G
溴三苯基甲烷 ≥98.0% (by titrimetric analysis)
溴三苯基甲烷
分子式:
(C₆H₅)₃CBr 沸点: 230 °C (1013 hPa) 密度: 1,55 g/cm³ (20 °C) |
MDL编号:
MFCD00000120 CAS号: 596-43-0 EINECS: 209-884-4 UN号: 3261 ADR: 8,III |
|
产品技术规格
Appearance | White to Yellow to Orange powder to crystal |
Purity (Argentmetric Titration) | min. 98.0 % |
Melting point | 153.0 to 157.0 deg-C |
Solubility in hot Acetic acid | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...