5-甲氧基-3-吲哚乙酸 ≥98.0% (by HPLC, titration analysis)
供应商: TCI
|
Warning
|
Synonyms:
5-Methoxy-3-indoleacetic acid, 2-(5-Methoxy-1H-indol-3-yl)acetic acid, 5-Methoxyindole-3-acetic acid
M1303-1G
M1303-5G
TCIAM1303-1GEA
0
CNY
TCIAM1303-1G
TCIAM1303-5G
5-甲氧基-3-吲哚乙酸 ≥98.0% (by HPLC, titration analysis)
5-甲氧基-3-吲哚乙酸
分子式:
C₁₁H₁₁NO₃ 分子量: 205.21 g/mol |
MDL编号:
MFCD00005638 CAS号: 3471-31-6 EINECS: 222-438-3 |
|
产品技术规格
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...