2-(三甲基硅基)乙醇 ≥96.0% (by GC)

供应商: TCI
Warning

Synonyms: (2-羟基乙基)三甲基硅烷

T1441-25ML T1441-5ML
TCIAT1441-25MLEA 0 CNY
TCIAT1441-25ML TCIAT1441-5ML
2-(三甲基硅基)乙醇 ≥96.0% (by GC)
2-(三甲基硅基)乙醇
分子式: HOH₂CCH₂Si(CH₃)₃
分子量: 118.25 g/mol
沸点: 75°C
溶点: 0
闪点: 50°C
MDL编号: MFCD00002825
CAS号: 2916-68-9
EINECS: 220-844-5
UN号: 1993
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8d326c638611a279dadfcf78b841b94ae0de0b47&vid=8d326c638611a279dadfcf78b841b94ae0de0b47&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR