1,1,2,2-四苯基-1,2-乙二醇 ≥95.0% (by HPLC)
供应商: TCI
|
Warning
|
Synonyms:
苯频哪醇
B1037-25G
B1037-5G
TCIAB1037-25GEA
0
CNY
TCIAB1037-25G
TCIAB1037-5G
1,1,2,2-四苯基-1,2-乙二醇 ≥95.0% (by HPLC)
1,1,2,2-四苯基-1,2-乙二醇
分子式:
(C₆H₅)₂C(OH)C(OH)(C₆H₅)₂ 分子量: 366.46 g/mol 沸点: 508 |
MDL编号:
MFCD00004448 CAS号: 464-72-2 EINECS: 207-356-8 UN号: 3077 ADR: 9 III Merck Index: 14,01100 |
|
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 95.0 area% |
Melting point | 178.0 to 184.0 deg-C |
Solubility in Chloroform | within almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...