(±)-1-Phenyl-1-pentanol ≥98.0% (by GC)
供应商: TCI
Synonyms: Fenipentol
P1086-25ML P1086-5ML
TCIAP1086-25MLEA 0 CNY
TCIAP1086-25ML TCIAP1086-5ML
(±)-1-Phenyl-1-pentanol ≥98.0% (by GC)
(±)-1-Phenyl-1-pentanol
分子式: C₆H₅CH(OH)(CH₂)₃CH₃
沸点: 137°C
MDL编号: MFCD00021935
CAS号: 583-03-9
EINECS: 209-493-9
Merck Index: 14,03975
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1affababfaeccf9be8a6641297c933131048efe1&vid=1affababfaeccf9be8a6641297c933131048efe1&prid=web&clid=SAPDAM
现在订购
产品技术规格
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %