1-(4-氟苯基)乙醇 ≥97.0% (by GC)

供应商: TCI
Warning

Synonyms: p-Fluoro-α-methylbenzyl alcohol, 1-(4-Fluorophenyl)ethyl Alcohol, 1-(4-Fluorophenyl)ethanol, 4-Fluorophenyl methyl carbinol

F0432-25G F0432-5G
TCIAF0432-25GEA 0 CNY
TCIAF0432-25G TCIAF0432-5G
1-(4-氟苯基)乙醇 ≥97.0% (by GC)
1-(4-氟苯基)乙醇
分子式: C₈H₉FO
分子量: 140.16 g/mol
沸点: 123°C
溶点: 9.3
密度: 1.1206
闪点: 91°C
MDL编号: MFCD00004515
CAS号: 403-41-8
EINECS: 206-959-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3108d02644106d30b9f31d1c1010bcef1e16c946&vid=3108d02644106d30b9f31d1c1010bcef1e16c946&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR