DL-脯氨酸 ≥99.0% (by titrimetric analysis)

供应商: TCI

Synonyms: (±)-吡咯烷-2-羧酸

P0480-1G P0480-25G
TCIAP0480-1GEA 0 CNY
TCIAP0480-1G TCIAP0480-25G
DL-脯氨酸 ≥99.0% (by titrimetric analysis)
DL-脯氨酸
分子式: C₅H₉NO₂
分子量: 115.13 g/mol
沸点: 245
MDL编号: MFCD00005250
CAS号: 609-36-9
EINECS: 210-189-3
Merck Index: 14,07780
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8325448b726b813f96a1e8e734dab4cd303b1298&vid=8325448b726b813f96a1e8e734dab4cd303b1298&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 99.0 %
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR