DL-赖氨酸一盐酸盐 ≥98.0% (by titrimetric analysis)
供应商: TCI
Synonyms:
DL-2,6-二氨基己酸
L0070-100G
L0070-25G
TCIAL0070-100GEA
0
CNY
TCIAL0070-100G
TCIAL0070-25G
DL-赖氨酸一盐酸盐 ≥98.0% (by titrimetric analysis)
DL-赖氨酸一盐酸盐
分子式:
C₆H₁₅ClN₂O₂ 分子量: 182.65 g/mol 溶点: 267 |
MDL编号:
MFCD00064563 CAS号: 70-53-1 EINECS: 200-739-0 |
|
产品技术规格
Appearance | White to Light yellow powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % |
Solubility in Water | very faint turbidity |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...