1,3-二甲基金刚烷 ≥99.0% (by GC)

供应商: TCI
Warning

D1081-25ML D1081-5ML
TCIAD1081-25MLEA 0 CNY
TCIAD1081-25ML TCIAD1081-5ML
1,3-二甲基金刚烷 ≥99.0% (by GC)
1,3-二甲基金刚烷
分子式: C₁₂H₂₀
沸点: 201°C
溶点: –30 °C
密度: 0,9016 g/cm³ (20 °C)
闪点: 53°C
MDL编号: MFCD00074755
CAS号: 702-79-4
EINECS: 211-870-8
UN号: 3295
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a467240d8e9a7b598829ee1398a7464d217e069&vid=9a467240d8e9a7b598829ee1398a7464d217e069&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR