2-异丁基-3-甲氧基吡嗪 ≥97.0% (by GC, titration analysis)
供应商: TCI
|
Warning
|
I0331-1ML
I0331-5ML
TCIAI0331-1MLEA
0
CNY
TCIAI0331-1ML
TCIAI0331-5ML
2-异丁基-3-甲氧基吡嗪 ≥97.0% (by GC, titration analysis)
2-异丁基-3-甲氧基吡嗪
分子式:
C₉H₁₄N₂O 沸点: 237 °C (1013 hPa) 溶点: 107 °C 密度: 0,99 g/cm³ (20 °C) 闪点: 80°C |
MDL编号:
MFCD00006128 CAS号: 24683-00-9 EINECS: 246-402-1 |
|
产品技术规格
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 97.0 % |
Purity (Nonaqueous Titration) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...