Dansyl chloride ≥98.0% (通过HPLC,滴定分析)
供应商: TCI
|
Danger
|
D0656-1G
D0656-25G
TCIAD0656-1GEA
0
CNY
TCIAD0656-1G
TCIAD0656-25G
Dansyl chloride ≥98.0% (通过HPLC,滴定分析)
Dansyl chloride
分子式:
C₁₂H₁₂ClNO₂S 溶点: 69…73 °C 存储条件: Refrigerator |
MDL编号:
MFCD00003985 CAS号: 605-65-2 EINECS: 210-092-6 UN号: 3261 ADR: 8,II Merck Index: 14,02814 |
|
产品技术规格
Appearance | Light yellow to Brown powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 68.0 to 73.0 deg-C |
Solubility in Acetone | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...