Di-tert-butyl sulphide ≥98.0% (by GC)

供应商: TCI
Warning

Synonyms: Di-tert-butyl sulphide, tert-Butyl sulphide

B0807-25ML B0807-500ML
TCIAB0807-25MLEA 0 CNY
TCIAB0807-25ML TCIAB0807-500ML
Di-tert-butyl sulphide ≥98.0% (by GC)
Di-tert-butyl sulphide
分子式: C₈H₁₈S
分子量: 146.3 g/mol
沸点: 150°C
闪点: 26°C
MDL编号: MFCD00008839
CAS号: 107-47-1
EINECS: 203-493-2
UN号: 1993
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0af8e0e23ba379323e8a29978de0a3b158b9eab1&vid=0af8e0e23ba379323e8a29978de0a3b158b9eab1&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR