二叔丁基二硫 ≥97.0% (by GC)
供应商: TCI
|
|
Synonyms:
叔丁基二硫
D0225-25ML
D0225-500ML
TCIAD0225-25MLEA
0
CNY
TCIAD0225-25ML
TCIAD0225-500ML
二叔丁基二硫 ≥97.0% (by GC)
二叔丁基二硫
分子式:
C₈H₁₈S₂ 沸点: 200°C 溶点: –5 °C 密度: 0,9229 g/cm³ (20 °C) 闪点: 67°C 存储条件: Ambient |
MDL编号:
MFCD00008838 CAS号: 110-06-5 EINECS: 203-734-1 UN号: 3082 ADR: 9,III |
|
产品技术规格
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...