4-氨基安替比林 99,0 GC, T

供应商: TCI
Warning

Synonyms: 氨基安替比林, 4-氨基-2,3-二甲基-1-苯基-3-吡唑啉-5-酮

A0256-25G A0256-500G
TCIAA0256-25GEA 0 CNY
TCIAA0256-25G TCIAA0256-500G
4-氨基安替比林 99,0 GC, T
4-氨基安替比林
分子式: C₁₁H₁₃N₃O
分子量: 203.24 g/mol
存储条件: Ambient
MDL编号: MFCD00003145
CAS号: 83-07-8
EINECS: 201-452-3
Merck Index: 14,00591
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3a94b41fb0d5a1f409308b793ef1610e86cfe4a1&vid=3a94b41fb0d5a1f409308b793ef1610e86cfe4a1&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Light yellow to Yellow to Orange powder to crystalline
Purity (GC) min. 99.0 %
Purity (Nonaqueous Titration) min. 99.0 %
Melting point 107.0 to 110.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR