1-对羟基苯基哌嗪 ≥98.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
4-(1-哌嗪基)苯酚
H0891-25G
H0891-250G
TCIAH0891-25GEA
0
CNY
TCIAH0891-25G
TCIAH0891-250G
1-对羟基苯基哌嗪 ≥98.0% (by GC)
1-对羟基苯基哌嗪
分子式:
C₁₀H₁₄N₂O 分子量: 178.23 g/mol |
MDL编号:
MFCD00066156 CAS号: 56621-48-8 EINECS: 260-289-6 |
|
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...