1-(3-甲氧基苯基)哌嗪 ≥98.0% (by GC, titration analysis)
供应商: TCI
|
Danger
|
M2135-25G
M2135-5G
TCIAM2135-25GEA
0
CNY
TCIAM2135-25G
TCIAM2135-5G
1-(3-甲氧基苯基)哌嗪 ≥98.0% (by GC, titration analysis)
1-(3-甲氧基苯基)哌嗪
分子式:
C₁₁H₁₆N₂O 沸点: 145°C 密度: 1,082 g/cm³ (20 °C) 闪点: 91°C |
MDL编号:
MFCD00040733 CAS号: 16015-71-7 EINECS: 240-154-8 UN号: 3267 ADR: 8,III |
|
产品技术规格
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...