3,4,5,6-四氯-1,2-苯醌 ≥97.0% (by HPLC, titration analysis)

供应商: TCI
Warning

Synonyms: 邻四氯苯醌

T0970-25G T0970-5G
TCIAT0970-25GEA 0 CNY
TCIAT0970-25G TCIAT0970-5G
3,4,5,6-四氯-1,2-苯醌 ≥97.0% (by HPLC, titration analysis)
3,4,5,6-四氯-1,2-苯醌
分子式: C₆Cl₄O₂
分子量: 245.88 g/mol
沸点: 312
MDL编号: MFCD00001646
CAS号: 2435-53-2
EINECS: 219-424-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8c9abdacfe17cf0c62f3a6d56626939c14b28e8c&vid=8c9abdacfe17cf0c62f3a6d56626939c14b28e8c&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Orange to Brown to Dark purple powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Iodometric Titration) min. 97.0 %
Melting point 126.0 to 130.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR