丙泊酚 ≥98.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
普鲁泊福, 2,6-二异丙基苯酚
D0617-25G
D0617-500G
TCIAD0617-25GEA
0
CNY
TCIAD0617-25G
TCIAD0617-500G
丙泊酚 ≥98.0% (by GC)
丙泊酚
分子式:
C₁₂H₁₈O 沸点: 256 °C (1013 hPa) 溶点: 18 °C 密度: 1,032 g/cm³ (20 °C) 闪点: 113°C |
MDL编号:
MFCD00008885 CAS号: 2078-54-8 EINECS: 218-206-6 UN号: 3082 ADR: 9,III Merck Index: 14,07834 |
|
产品技术规格
Appearance | Colorless to Red to Green clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...