2,2,2-三苯基苯乙酮 ≥99.0% (通过GC)
供应商: TCI
T1207-25G
T1207-5G
TCIAT1207-25GEA
0
CNY
TCIAT1207-25G
TCIAT1207-5G
2,2,2-三苯基苯乙酮 ≥99.0% (通过GC)
2,2,2-三苯基苯乙酮
分子式:
C₂₆H₂₀O 沸点: 498 °C (1013 hPa) 溶点: 178 °C |
MDL编号:
MFCD00004762 CAS号: 466-37-5 EINECS: 207-375-1 |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 99.0 % |
Melting point | 182.0 to 185.0 deg-C |
Solubility in hot Toluene | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...