二苯乙腈 ≥99.0% (by GC)

供应商: TCI
Warning

Synonyms: 2,2-Diphenylacetonitrile

D0261-25G D0261-500G
TCIAD0261-25GEA 0 CNY
TCIAD0261-25G TCIAD0261-500G
二苯乙腈 ≥99.0% (by GC)
二苯乙腈
分子式: (C₆H₅)₂CHCN
沸点: 181 °C (1013 hPa)
溶点: 71…74 °C
密度: 1,182 g/cm³ (20 °C)
存储条件: Ambient
MDL编号: MFCD00001862
CAS号: 86-29-3
EINECS: 201-662-5
Merck Index: 14,03316
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=94032d94a87ac21acdf5e0d07b286cab07800d49&vid=94032d94a87ac21acdf5e0d07b286cab07800d49&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Light yellow powder to crystal
Purity (GC) min. 99.0 %
Melting point 73.0 to 76.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR