4-Hexylphenol ≥98.0% (by GC)
供应商: TCI
|
Danger
|
H0611-25G
H0611-5G
TCIAH0611-25GEA
0
CNY
TCIAH0611-25G
TCIAH0611-5G
4-Hexylphenol ≥98.0% (by GC)
4-Hexylphenol
分子式:
C₁₂H₁₈O 沸点: 100 °C (0,01 -rr) 溶点: 28 °C 密度: 0,9628 g/cm³ (20 °C) |
MDL编号:
MFCD00051313 CAS号: 2446-69-7 EINECS: 219-501-2 UN号: 2430 ADR: 8,III |
产品技术规格
Appearance | White or Colorless to Light yellow powder to lump to clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...