4'-氟-2'-羟基苯乙酮 ≥97.0% (by GC)

供应商: TCI
Warning

Synonyms: 4-Fluoro-2-hydroxyacetophenone

F0723-25G F0723-5G
TCIAF0723-25GEA 0 CNY
TCIAF0723-25G TCIAF0723-5G
4'-氟-2'-羟基苯乙酮 ≥97.0% (by GC)
4'-氟-2'-羟基苯乙酮
分子式: C₈H₇FO₂
分子量: 154.14 g/mol
MDL编号: MFCD00143203
CAS号: 1481-27-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6193a715028c5e66899386783e03f63f62cd2146&vid=6193a715028c5e66899386783e03f63f62cd2146&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to lump
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR