苯基溴化硒 ≥97.0% (by titrimetric analysis)

供应商: TCI
Danger

Synonyms: Benzeneselenyl bromide, 苯硒基溴化物

P1322-25G P1322-5G
TCIAP1322-25GEA 0 CNY
TCIAP1322-25G TCIAP1322-5G
苯基溴化硒 ≥97.0% (by titrimetric analysis)
苯基溴化硒
分子式: (C₆H₅)SeBr
分子量: 235.97 g/mol
沸点: 107°C
MDL编号: MFCD00000047
CAS号: 34837-55-3
EINECS: 252-238-1
UN号: 2928
ADR: 6.1 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e394b3769ed10e67e4ede57e130b88f54fdffbbf&vid=e394b3769ed10e67e4ede57e130b88f54fdffbbf&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Orange to Brown to Dark purple powder to crystal
Purity (Iodometric Titration) min. 97.0 %
Melting point 56.0 to 60.0 deg-C
Solubility in Toluene very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR