2-甲氧基-4-硝基苯胺 ≥99.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
4-硝基邻茴香胺
M0118-25G
M0118-500G
TCIAM0118-25GEA
0
CNY
TCIAM0118-25G
TCIAM0118-500G
2-甲氧基-4-硝基苯胺 ≥99.0% (by GC)
2-甲氧基-4-硝基苯胺
分子式:
C₇H₈N₂O₃ 分子量: 168.15 g/mol 沸点: 364 溶点: 139 存储条件: Ambient |
MDL编号:
MFCD00007363 CAS号: 97-52-9 EINECS: 202-588-6 |
|
产品技术规格
Appearance | Light yellow to Brown powder to crystal |
Purity (GC) | min. 99.0 % |
Melting point | 139.0 to 143.0 deg-C |
Solubility in hot Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...