2-硝基-4-氟苯酚 ≥98.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
1-Fluoro-4-hydroxy-3-nitrobenzene
F0367-25G
F0367-5G
TCIAF0367-25GEA
0
CNY
TCIAF0367-25G
TCIAF0367-5G
2-硝基-4-氟苯酚 ≥98.0% (by GC)
2-硝基-4-氟苯酚
分子式:
C₆H₄FNO₃ 分子量: 157.1 g/mol 溶点: 74 |
MDL编号:
MFCD00042528 CAS号: 394-33-2 |
|
产品技术规格
Appearance | Orange to Green to Amber powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 74.0 to 77.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...