5-硝基间苯二甲酸二甲酯

供应商: TCI

Synonyms: 5-硝基苯-1,3-二羧酸二甲酯

N0567-25G N0567-500G
TCIAN0567-25GEA 0 CNY
TCIAN0567-25G TCIAN0567-500G
5-硝基间苯二甲酸二甲酯
5-硝基间苯二甲酸二甲酯
分子式: C₁₀H₉NO₆
MDL编号: MFCD00008429
CAS号: 13290-96-5
EINECS: 236-307-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=af34769c1a38c6fd269803cc26c820ec1fe7d8d8&vid=af34769c1a38c6fd269803cc26c820ec1fe7d8d8&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 123.0 to 126.0 deg-C
Solubility in Toluene almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR