3-Chloro-4-iodoaniline ≥95.0% (by GC)

供应商: TCI
Warning

C2167-25G C2167-5G
TCIAC2167-25GEA 0 CNY
TCIAC2167-25G TCIAC2167-5G
3-Chloro-4-iodoaniline ≥95.0% (by GC)
3-Chloro-4-iodoaniline
分子式: C₆H₅ClIN
分子量: 253.47 g/mol
沸点: 289
溶点: 65
MDL编号: MFCD00051845
CAS号: 135050-44-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8474fc39633ca1ec055eb9a432c5e4c8b3a2ec49&vid=8474fc39633ca1ec055eb9a432c5e4c8b3a2ec49&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Amber to Dark purple powder to crystal
Purity (GC) min. 95.0 %
Melting point 68.0 to 72.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR