2,6-二氯-4-(三氟甲氧基)苯胺 ≥98.0% (by GC)
供应商: TCI
|
Warning
|
D3802-25G
D3802-5G
TCIAD3802-25GEA
0
CNY
TCIAD3802-25G
TCIAD3802-5G
2,6-二氯-4-(三氟甲氧基)苯胺 ≥98.0% (by GC)
2,6-二氯-4-(三氟甲氧基)苯胺
分子式:
C₇H₄Cl₂F₃NO 分子量: 246.02 g/mol |
MDL编号:
MFCD00190128 CAS号: 99479-66-0 |
|
产品技术规格
Appearance | White to Gray to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 30.0 to 34.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...