4-溴-2-(三氟甲氧基)苯胺 ≥98.0% (by GC)

供应商: TCI
Warning

B3786-25G B3786-5G
TCIAB3786-25GEA 0 CNY
TCIAB3786-25G TCIAB3786-5G
4-溴-2-(三氟甲氧基)苯胺 ≥98.0% (by GC)
4-溴-2-(三氟甲氧基)苯胺
分子式: C₇H₅BrF₃NO
分子量: 256.02 g/mol
沸点: 115°C
MDL编号: MFCD00179338
CAS号: 175278-09-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3c90516275c07be70e87f775b74dc9680da67497&vid=3c90516275c07be70e87f775b74dc9680da67497&prid=web&clid=SAPDAM

现在订购

产品技术规格

Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR