γ-亚麻酸 ≥98.0% (by GC, titration analysis)
供应商: TCI
|
Warning
|
Synonyms:
顺,顺,顺-6,9,12-十八碳三烯酸
L0152-1G
L0152-100MG
TCIAL0152-1GEA
0
CNY
TCIAL0152-1G
TCIAL0152-100MG
γ-亚麻酸 ≥98.0% (by GC, titration analysis)
γ-亚麻酸
分子式:
C₁₈H₃₀O₂ 分子量: 278.44 g/mol 存储条件: Refrigerator |
MDL编号:
MFCD00065718 CAS号: 506-26-3 Merck Index: 14,05507 |
|
产品技术规格
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...