频哪醇 ≥98.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
2,3-二甲基-2,3-丁二醇, 四甲基乙二醇
D0691-25G
D0691-500G
TCIAD0691-25GEA
0
CNY
TCIAD0691-25G
TCIAD0691-500G
频哪醇 ≥98.0% (by GC)
频哪醇
分子式:
C₆H₁₄O₂ 分子量: 118.18 g/mol 沸点: 174 闪点: 77 |
MDL编号:
MFCD00004462 CAS号: 76-09-5 EINECS: 200-933-5 Merck Index: 14,07439 |
|
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 41.0 to 45.0 deg-C |
Solubility in Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...