正丁基丙二酸二乙酯 ≥99.0% (by GC)
供应商: TCI
Synonyms:
Butylmalonic acid diethyl ester, Butyl-1,3-propanedioic acid diethyl ester, Diethyl 2-butylmalonate
B0381-25ML
B0381-500ML
TCIAB0381-25MLEA
0
CNY
TCIAB0381-25ML
TCIAB0381-500ML
正丁基丙二酸二乙酯 ≥99.0% (by GC)
正丁基丙二酸二乙酯
分子式:
C₁₁H₂₀O₄ 沸点: 240°C 密度: 0,976 g/cm³ (20 °C) 闪点: 94°C |
MDL编号:
MFCD00009170 CAS号: 133-08-4 EINECS: 205-089-1 Merck Index: 13,01575 |
|
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...